missing translation for 'onlineSavingsMsg'
Learn More

Ergodyne™ Chill-Its™ 6665 Evaporative Cooling Vest

Catalog No. 19910202
Change view
Click to view available options
Color:
Gray
Lime
Size:
Medium
Large
X-Large
2X-Large
3X-Large
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Size
19910202 Lime Medium
19910203 Lime Large
19910204 Lime X-Large
19910205 Lime 2X-Large
19910206 Gray Medium
19910207 Gray Large
19910208 Gray X-Large
19910209 Gray 3X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 19910202 Supplier Ergodyne™ Supplier No. 12533

Evaporative Cooling Vest provides cooling relief and comfort once soaked in cold water.

  • V-neck with zipper closure
  • Quilted nylon outer layer
  • Activated, polymer-embedded fabric inside
  • Water-repellent liner and black cotton-poly elastic trim
  • Lightweight, durable, and washable
  • Soak in water 2-5 minutes to activate
  • Remains hydrated for up to 4 hours
  • Mesh side panels offer improved ventilation and stretch

Specifications

Material Nylon
Product Type Vest
Size Medium
Color Lime
For Use With (Application) Construction, Maintenance, Landscaping, Material Handling