missing translation for 'onlineSavingsMsg'
Learn More

Ergodyne™ Chill-Its™ 6660 Hard Hat Brim with Shade

Catalog No. p-7163092
Change view
Click to view available options
Color:
Lime
Orange
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19819601 Lime
19819602 Orange
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19819601 Supplier Ergodyne™ Supplier No. 12640
Only null left

Hard Hat Brim with Shade fits around the outside of a hard hat to provide sun and UV protection on the face and neck.

  • 100% Polyester fabric
  • Universal design
  • Elastic inner rim to fit most hard hats
  • Designed to block sun and keep workers comfortable in outdoor environments

Specifications

Color Lime
For Use With (Application) Construction, Service, Landscaping, Material Handling
Material Polyester