missing translation for 'onlineSavingsMsg'
Learn More

Ergodyne™ Chill-Its™ 6650 High Performance Hat with Neck Shade

Catalog No. 19819542
Change view
Click to view available options
Color:
Lime
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19819542 Lime
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19819542 Supplier Ergodyne™ Supplier No. 12520

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

High-Performance Hat with Neck Shade is a machine washable hat designed to get moisture away from skin.

  • Hi Cool™ moisture management performance fabric
  • Absorbent terry headband
  • Wide panel neck protection
  • Adjustable cinch cord for a great fit

Specifications

Material Hi Cool™ Moisture Management Performance Fabric
Product Type Headband
Color Lime
Closure Type Adjustable Cinch Cord
For Use With (Application) Construction, Maintenance, Landscaping, Distribution