missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ergodyne™ Chill-Its™ 6630 High-Performance Cap
Description
- Hi Cool™ Moisture Management performance fabric
- Comfortable to wear under hats, helmets, or alone
- Wide, absorbent terrycloth band
- Elastic band for a great fit
Specifications
Specifications
| Material | Fabric |
| Color | Hi-vis Lime |
| For Use With (Application) | Construction, trades, maintenance, landscaping/grounds, assembly/fabrication, material handling, freight/baggage, warehousing/distribution, Iron/Steel fabrication, hells kitchen |
| For Use With (Equipment) | For use under hats, helmets, or alone |
| Includes | Elastic Band |