missing translation for 'onlineSavingsMsg'
Learn More

Ergodyne™ Chill-Its™ 6615 High-Performance Dew Rag

Catalog No. p-7163089
Change view
Click to view available options
Color:
Black
Gray, Black, White
Lime
Realtree Xtra™
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19819523 Realtree Xtra™
19819524 Lime
19819531 Black
19819540 Gray, Black, White
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 19819523 Supplier Ergodyne™ Supplier No. 12475
Only null left

High-Performance Dew Rag features technical fabric to keep away moisture and an elastic band for a secure fit.

  • Hi Cool™ moisture management performance fabric
  • Comfortable to wear under hats, helmets, or alone
  • Absorbent terry headband
  • Elastic band for a great fit
  • Available in a variety of patterns and colors like black, lime, skulls, and Realtree Xtra™

Specifications

Product Type Dew Rag
Color Realtree Xtra™
Band Type Elastic
For Use With (Application) Construction, Maintenance, Landscaping