missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ergodyne™ Chill-Its™ 6615 High-Performance Dew Rag
Description
- Hi Cool™ moisture management performance fabric
- Comfortable to wear under hats, helmets, or alone
- Absorbent terry headband
- Elastic band for a great fit
- Available in a variety of patterns and colors like black, lime, skulls, and Realtree Xtra™
Specifications
Specifications
| Product Type | Dew Rag |
| Color | Realtree Xtra™ |
| For Use With (Application) | Construction, Maintenance, Landscaping |