missing translation for 'onlineSavingsMsg'
Learn More

Ergodyne™ Chill-Its™ 6605 High-Performance Headband

Catalog No. p-7163088
Change view
Click to view available options
Color:
Black
Blue
Camouflage
Red, White, Black
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19819509 Camouflage
19819510 Red, White, Black
19819513 Blue
19819516 -
19819517 Black
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 19819509 Supplier Ergodyne™ Supplier No. 12422
Only null left

High-Performance Headband constructed of technical fabric to absorb moisture away from the body for fast evaporation.

  • Hi Cool™ moisture management performance fabric
  • Absorbent terry headband
  • Elasticized back for a great fit
  • Available in the following patterns: Stars and Stripes, Camouflage, Red Western, Navy Western, Blue, Lime, Flames, and Skulls

Specifications

Material Hi Cool™ Moisture Management Performance Fabric
Product Type Headband
Color Camouflage
For Use With (Application) Construction, Maintenance, Landscaping