missing translation for 'onlineSavingsMsg'
Learn More

Ergodyne™ Chill-Its™ 6603 Evaporative Cooling Band

Catalog No. 19819640
Change view
Click to view available options
Color:
Blue
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19819640 Blue
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19819640 Supplier Ergodyne™ Supplier No. 12418

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chill-Its™ 6603 Evaporative Cooling Band is designed to absorb water and create a feeling colder than ambient air temperature.

  • Instant cooling relief
  • Run under water for one minute or until saturated, wring out excess water for drip free cooling
  • Pull the end through the slit for a comfortable and secure fit around neck
  • Can last for up to 4 hours on a single soak
  • Cooling PVA material absorbs water and creates a cooling sensation as it evaporates
  • Reusable
  • Machine washable with mild detergent, air dry only
  • Reduces the effect of heat exhaustion and fights fatigue while working or playing in the heat

Specifications

Material PVA
Color Blue
Dimensions (L x W) 4 x 29.5 in. (10.2 x 74.9 cm)
For Use With (Application) Construction, Landscaping