missing translation for 'onlineSavingsMsg'
Learn More

Ergodyne™ Chill-Its™ 6602 Evaporative Cooling Towel

Catalog No. p-7163086
Change view
missing translation for 'orderingAttributeHoverText'
Color:
Gray
Orange
Pink
Catalog No. Color
19819644 Gray
19819648 Orange
19819649 Pink
3 options

Catalog No. 19819644

Supplier: Ergodyne™ 12438

Only null left

Chill-Its™ 6602 Evaporative Cooling Towel uses a PVA material to absorb water to allow slow evaporation for instant, long-lasting cooling relief.

  • Run under water for one minute or until saturated, wring out excess water for drip free cooling
  • Can last for up to 4 hours on a single soak. Simply re-wet to increase cooling duration
  • Cooling PVA material absorbs water and creates a cooling sensation as it evaporates
  • Air dry until material is stiff, re-wet until soft
  • Machine washable with mild detergent, air dry only
  • Reduces the effects of heat exhaustion and fights fatigue

Specifications

Material PVA
Product Type Cooling Towel
Color Gray
Dimensions (L x W) 13 x 29.5 in. (33 x 75 cm)
For Use With (Application) Construction, Agriculture, Landscaping