missing translation for 'onlineSavingsMsg'
Learn More

Ergodyne™ Chill-Its™ 6602 Evaporative Cooling Towel

Catalog No. 19819649
Change view
Click to view available options
Color:
Gray
Orange
Pink
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19819649 Pink
19819644 Gray
19819648 Orange
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 19819649 Supplier Ergodyne™ Supplier No. 12442

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chill-Its™ 6602 Evaporative Cooling Towel uses a PVA material to absorb water to allow slow evaporation for instant, long-lasting cooling relief.

  • Run under water for one minute or until saturated, wring out excess water for drip free cooling
  • Can last for up to 4 hours on a single soak. Simply re-wet to increase cooling duration
  • Cooling PVA material absorbs water and creates a cooling sensation as it evaporates
  • Air dry until material is stiff, re-wet until soft
  • Machine washable with mild detergent, air dry only
  • Reduces the effects of heat exhaustion and fights fatigue

Specifications

Material PVA
Product Type Cooling Towel
Color Pink
Dimensions (L x W) 13 x 29.5 in. (33 x 75 cm)
For Use With (Application) Construction, Agriculture, Landscaping