missing translation for 'onlineSavingsMsg'
Learn More

Ergodyne™ Chill-Its™ 6225 Premium FR Phase Change Cooling Vest

Catalog No. p-7163080
Change view
Click to view available options
Color:
Orange
Tan
Size:
Large/X-Large
Small/Medium
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Size
19819676 Tan Small/Medium
19819682 Tan Large/X-Large
19819684 Orange Large/X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 19819676 Supplier Ergodyne™ Supplier No. 12203

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chill-Its™ 6215 Premium Phase Change Cooling Vest keeps workers cool and safe when combined with a cooling pack.

  • Wash by hand with a mild detergent and warm water
  • Flame resistant
  • Made with modacrylic cotton blend FR fabric
  • Adjustable hook and loop closures on both shoulders and waist
  • Extended vest surface area enhances cooling power
  • Designed for more visibility on busy streets or job sites
  • Two elastic pen loops on chest

Specifications

Material Modacrylic Cotton Blend FR Fabric
Product Type Cooling Vest
Size Small/Medium
Length (Metric) 81 to 102 cm
Length (English) 32 to 40 in.
Color Tan
Certifications/Compliance ASTM F1506 performance requirements (ATPV rating of 7.1 cal/cm2); Classified to NFPA 70E (CAT1)
For Use With (Application) Fire Rescue, EMT, Construction
Quantity 6/Pk.