missing translation for 'onlineSavingsMsg'
Learn More

Ergodyne™ Chill-Its™ 6220 Phase Change Cooling Vest Charge Packs

Catalog No. 19819671
Change view
Click to view available options
Size:
Large/X-Large
Small/Medium
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19819671 Small/Medium
19819672 Large/X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19819671 Supplier Ergodyne™ Supplier No. 12201

Chill-Its™ 6220 Phase Change Cooling Vest Charge Packs are suitable for use with a variety of cooling vests.

  • Immediate and long-lasting cooling relief
  • Packs maintain constant 64°F (18°C) for up to 4 hours
  • Quick recharge in a little as 10 minutes
  • Patented honeycomb ice pack design eliminates stiffness
  • Wash by hand with a mild detergent and warm water

Specifications

Material Cotton with Gel Polymer
Product Type Cooling Pack
Size Small/Medium
Color Clear
Cooling Time Packs Maintain Constant 64°F (18°C) for up to 4 hours
For Use With (Application) Cooling Vest
Quantity 6/Pk.