missing translation for 'onlineSavingsMsg'
Learn More

Ergodyne™ Chill-Its™ 5159 Hydration Pack Cleaning Kit

Catalog No. 19819780
Change view
Click to view available options
Color:
Blue
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19819780 Blue
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19819780 Supplier Ergodyne™ Supplier No. 13159

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chill-Its™ 5159 Hydration Pack Cleaning Kit is designed to help keep all the components of a hydration pack clean and fresh.

  • Compatible with any Chill-Its™ bladder
  • Drying rack to air dry bladder
  • Bladder cleaning brush
  • Delivery system cleaning brush
  • Bite-valve cleaning brush

Specifications

Color Blue
For Use With (Application) Cleaning hydration pack