missing translation for 'onlineSavingsMsg'
Learn More

Ergodyne™ Chill-Its™ 5051B Bladder Replacement

Catalog No. 19819750
Change view
Click to view available options
Capacity:
3 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity
19819750 3 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19819750 Supplier Ergodyne™ Supplier No. 13059

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chill-Its™ 5051B Bladder Replacement is designed for use with premium low profile hydration packs and premium cargo hydration packs.

  • TPE bladder eliminates plastic odor and taste
  • Stays supple to withstand daily wear and tear
  • Dual cap bladder featuring smaller (60 mm) cap nestled into larger (80 mm) cap for easy water and ice filling
  • 100% anti-microbial bite valve with cover to protect against contaminants
  • Insulated tube keeps water cool

Specifications

Color Blue
Capacity 3 L
For Use With (Application) Fire, Rescue, EMT, Construction, Landscaping