missing translation for 'onlineSavingsMsg'
Learn More

Ergodyne™ Chill-Its™ 5050M Mouthpiece Replacement

Catalog No. 19819749
Change view
Click to view available options
Color:
Black
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19819749 Black
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19819749 Supplier Ergodyne™ Supplier No. 13058

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chill-Its™ 5050M Mouthpiece Replacement is designed for use with hydration packs.

  • Replacement bite valve
  • For use with hydration packs

Specifications

Color Black
For Use With (Application) Staying hydrated, hiking