missing translation for 'onlineSavingsMsg'
Learn More

Ergodyne™ Chill-Its™ 5050B Bladder Replacement

Catalog No. 19819748
Change view
Click to view available options
Capacity:
2 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity
19819748 2 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19819748 Supplier Ergodyne™ Supplier No. 13057

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chill-Its™ 5050B Bladder Replacement is designed for use with a low profile hydration pack.

  • Wide mouth opening for easy water and ice refilling
  • Insulated tube keeps water cool

Specifications

Color Blue
Capacity 2 L
For Use With (Application) Staying hydrated, hiking