missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chicoric Acid 98.0+%, TCI America™
Supplier: TCI America C345050MG
Specifications
| Chicoric Acid | |
| 206°C | |
| MFCD22683653 | |
| (2R,3R)-2,3-Bis[[(E)-3-(3,4-dihydroxyphenyl)acryloyl]oxy]succinic Acid | |
| C1=CC(=C(C=C1C=CC(=O)OC(C(C(=O)O)OC(=O)C=CC2=CC(=C(C=C2)O)O)C(=O)O)O)O | |
| 474.374 | |
| CHEBI:3594 | |
| ≥98.0% (T) |
| 70831-56-0 | |
| C22H18O12 | |
| 50 mg | |
| YDDGKXBLOXEEMN-IABMMNSOSA-N | |
| (2R,3R)-2,3-bis[[(E)-3-(3,4-dihydroxyphenyl)prop-2-enoyl]oxy]butanedioic acid | |
| 5281764 | |
| 474.37 | |
| Crystalline Powder |
Chemical Identifiers
| 70831-56-0 | |
| 474.374 | |
| YDDGKXBLOXEEMN-IABMMNSOSA-N | |
| 5281764 | |
| (2R,3R)-2,3-bis[[(E)-3-(3,4-dihydroxyphenyl)prop-2-enoyl]oxy]butanedioic acid |
| C22H18O12 | |
| MFCD22683653 | |
| (2R,3R)-2,3-Bis[[(E)-3-(3,4-dihydroxyphenyl)acryloyl]oxy]succinic Acid | |
| CHEBI:3594 | |
| C1=CC(=C(C=C1C=CC(=O)OC(C(C(=O)O)OC(=O)C=CC2=CC(=C(C=C2)O)O)C(=O)O)O)O |
Safety and Handling
RTECSNumber : EJ9852090
TSCA : No
Recommended Storage : Refrigerator