missing translation for 'onlineSavingsMsg'
Learn More

Chicoric Acid 98.0+%, TCI America™

Catalog No. C345010MG
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C345010MG 10 mg
C345050MG 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C345010MG Supplier TCI America Supplier No. C345010MG
Only null left

Chemical Identifiers

CAS 70831-56-0
Molecular Formula C22H18O12
Molecular Weight (g/mol) 474.374
MDL Number MFCD22683653
InChI Key YDDGKXBLOXEEMN-IABMMNSOSA-N
Synonym (2R,3R)-2,3-Bis[[(E)-3-(3,4-dihydroxyphenyl)acryloyl]oxy]succinic Acid
PubChem CID 5281764
ChEBI CHEBI:3594
IUPAC Name (2R,3R)-2,3-bis[[(E)-3-(3,4-dihydroxyphenyl)prop-2-enoyl]oxy]butanedioic acid
SMILES C1=CC(=C(C=C1C=CC(=O)OC(C(C(=O)O)OC(=O)C=CC2=CC(=C(C=C2)O)O)C(=O)O)O)O

Specifications

Melting Point 206°C
Quantity 10 mg
Formula Weight 474.37
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Chicoric Acid
RTECSNumber EJ9852090
TSCA No
Recommended Storage Refrigerator