Learn More
Thermo Scientific™ Chest Freezer Racks
Use Thermo Scientific™ Chest Freezer Racks with Forma™, HERAfreeze™, Revco™ and TS-Series Ultra-Low Temperature Chest Freezers.
Supplier:  Thermo Scientific™ 398184
 View more versions of this product
Catalog No. 398184
Only null left
Description
Description
Use Thermo Scientific™ Chest Freezer Racks with TDE Series Ultra-Low Temperature Chest Freezers as well as legacy Forma™, HERAfreeze™, Revco™ and TS-Series Ultra-Low Temperature Chest Freezers.
  • Warranty varies by country, please contact us for details
Specifications
Chemical Identifiers
Silver
Specifications
Silver
Stainless Steel
3 cu. ft. Chest Freezers
5.5 in.
5.5 in.
Side Access Rack Tower
14 cm
Boxes (2 in.)
7 Boxes/Rack
3 cu. ft.
15.2 in.
14 cm
38.6 cm
Product Suggestions
Product Suggestions