Learn More
Thermo Scientific™ Chest Freezer Racks
Use Thermo Scientific™ Chest Freezer Racks with Forma™, HERAfreeze™, Revco™ and TS-Series Ultra-Low Temperature Chest Freezers.
Supplier:  Thermo Scientific™ 398188
 View more versions of this product
Catalog No. 13990478
Only null left
Description
Description
Use Thermo Scientific™ Chest Freezer Racks with TDE Series Ultra-Low Temperature Chest Freezers as well as legacy Forma™, HERAfreeze™, Revco™ and TS-Series Ultra-Low Temperature Chest Freezers.
  • Warranty varies by country, please contact us for details
Specifications
Chemical Identifiers
Silver
Specifications
Silver
27
12.7, 17, 20 cu. ft. Freezers
5.3 in.
3.5 in.
Microplate Rack
13.5 cm
Plates
Rectangular
12.7, 17, 20  cu. ft.
25.8 in.
8.9 cm
65.5 cm
Product Suggestions
Product Suggestions