missing translation for 'onlineSavingsMsg'
Learn More
Learn More
International Enviroguard™ ChemSplash 1 Chemical Splash Sleeves with Serged Seams and Elastic Closures
Description
- Proprietary, hybrid membrane fabric helps keep arms cool as you work
- Secure, elastic closures on both ends help keep sleeves in place with repetitive movements
- Longer length provides more arm coverage to prevent skin or underlying clothing from exposure to workplace hazards
- Elastic closures on both ends of sleeves offer a secure fit to keep sleeves in place to prevent skin exposure
- High moisture vapor transfer rate reduces heat and moisture build-up to keep wearers cooler
- Possible applications include manufacturing, emergency response, environmental cleanup, and more
Specifications
Specifications
| Size | Universal |
| Product Type | Chemical-resistant Sleeve |
| Material | Non-woven Fabric |
| Color | Yellow |
| Sterility | Non-sterile |
| Style | Elastic at Both Ends |
| Disposable | Single-use |
| Seam Type | Serged |
| Latex Free | Yes |
| Length (English) | 18 in. |
| Show More |