missing translation for 'onlineSavingsMsg'
Learn More
Learn More
International Enviroguard™ ChemSplash 1 Chemical Splash Coveralls with Serged Seams, Elastic Wrists and Open Ankles
These durable and disposable chemical coveralls with serged seams and elastic back protect against chemical splash and toxic particles.
Supplier: International Enviroguard™ 7012YS2XL
Description
- Durable and disposable chemical coveralls with serged seams and elastic back protect against chemical splash and toxic particles
- Proprietary, hybrid membrane fabric offers a high moisture vapor transfer rate (MVTR), so you stay cool as you put in a hard day's work
- A storm flap over the zipper helps prevent hazardous liquids from seeping through otherwise exposed zipper seams
- For added comfort, seamless shoulder areas help prevent tears in high-movement areas and allow you to move more freely as you work
- ChemSplash chemical suits also pass ASTM F1670 testing for blood penetration
- Chemical protective coveralls are designed with serged seams with possible applications including manufacturing, emergency response, environmental cleanup, and more
- A wider, gusseted crotch gives the wearer more room to move, bend, crouch, and climb without ripping fabric
- Elastic back stretches with user for more comfort and better tear resistance
- High moisture vapor transfer rate (MVTR) doesn't trap heat and moisture near the body, so workers stay cooler to prevent heat stress
- Elastic wrists offer a secure fit that keeps sleeves in place to help prevent skin exposure
Specifications
| Chemical Protective Clothing | |
| Unisex | |
| Yellow | |
| Elastic at Back | |
| Elastic | |
| Standard Collar | |
| Serged | |
| Chemicals, Particle, Caustics, Acid, Isopropyl Alcohol | |
| None |
| 2X-Large | |
| Laminated Non-woven Fabric | |
| Single-use | |
| Open | |
| Collared | |
| 2-way Front Zipper with Storm Flap | |
| Non-sterile | |
| Yes |