missing translation for 'onlineSavingsMsg'
Learn More
Learn More
International Enviroguard™ ChemSplash 1 Chemical Splash Coveralls with Serged Seams, Elastic Wrists and Open Ankles
Description
- Durable and disposable chemical coveralls with serged seams and elastic back protect against chemical splash and toxic particles
- Proprietary, hybrid membrane fabric offers a high moisture vapor transfer rate (MVTR), so you stay cool as you put in a hard day's work
- A storm flap over the zipper helps prevent hazardous liquids from seeping through otherwise exposed zipper seams
- For added comfort, seamless shoulder areas help prevent tears in high-movement areas and allow you to move more freely as you work
- ChemSplash chemical suits also pass ASTM F1670 testing for blood penetration
- Chemical protective coveralls are designed with serged seams with possible applications including manufacturing, emergency response, environmental cleanup, and more
- A wider, gusseted crotch gives the wearer more room to move, bend, crouch, and climb without ripping fabric
- Elastic back stretches with user for more comfort and better tear resistance
- High moisture vapor transfer rate (MVTR) doesn't trap heat and moisture near the body, so workers stay cooler to prevent heat stress
- Elastic wrists offer a secure fit that keeps sleeves in place to help prevent skin exposure
Specifications
Specifications
| Product Type | Chemical Protective Clothing |
| Size | Large |
| Gender | Unisex |
| Material | Laminated Non-woven Fabric |
| Color | Yellow |
| Disposable | Single-use |
| Waist Style | Elastic at Back |
| Ankle Style | Open |
| Cuff Style | Elastic |
| Neck Style | Collared |
| Show More |