missing translation for 'onlineSavingsMsg'
Learn More

Azure Biosystems ChemiWriter ECL Pen

Catalog No. AC2146AZ
Change view
Click to view available options
Type:
Chemiluminescent Pen
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
AC2146AZ Chemiluminescent Pen
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC2146AZ Supplier Azure Biosystems Supplier No. AC2146

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemiluminescent marker that enables to write or draw on transfer membranes, resulting in a chemiluminescent signal on markings once substrate is added, 1 piece

  • Chemiluminescent pen for blot annotation
  • The Azure ChemiWriter ECL Western blot annotation pen is a chemiluminescent marker that enables you to write or draw on your transfer membranes, resulting in a chemiluminescent signal on your markings once substrate is added. Contains a reagent that reacts with HRP substrates to produce chemiluminescence and also a blue dye to allow you to see markings as you write on the Western blot.
  • Use for tracing visible protein ladders or making identifying marks on your Western blots. Pen has two tips on each end: one fine-tipped for thin markings and one thicker for depositing more of the reagent.
  • Quantity: 1 pen

Specifications

Type Chemiluminescent Pen
Format Liquid
For Use With Western Blot