missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Chemicon™ Neural Stem Cell Marker Characterization Kit

Catalog No. SCR019
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SCR019 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SCR019 Supplier MilliporeSigma™ Supplier No. SCR019

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Contains two molecular markers, Nestin and Sox 2 that are frequently used to identify neural stem/progenitor cells.

  • Applications: Immunocytochemistry, Stem Cell Culture

Specifications

Cell Type Neural Stem Cells
Product Type Stem Cell Characterization Kit
For Use With (Application) Immunocytochemistry, Stem Cell Culture
Quantity 1 Kit
Regulatory Status RUO
Content And Storage
  • Mouse anti-Nestin: (Catalog No. 2003602) One vial containing 50μg monoclonal antibody. Store at 2° to 8°C.
  • Rabbit anti-Sox 2: (Catalog No. 2003600) One vial containing 20μg affinity purified polyclonal antibody. Store at 2° to 8°C.
  • Mouse anti-MAP-2: (Catalog No. MAB3418-50UG) One vial containing 50μg monoclonal antibody. Store at 2° to 8°C.
  • Rabbit anti-GFAP: (Catalog No. AB5804) One vial containing 50μL of polyclonal rabbit serum. Store at -20°C.
  • Mouse anti-Oligodendrocyte marker O1: (Catalog No. 2003601) One vial containing 20μg IgM monoclonal antibody. Store at -20°C.
  • Mouse IgM: (Catalog No. 2003599) One vial containing 50μg purified mouse IgM control antibody. Store at 2° to 8°C.
  • Mouse IgG: (Catalog No. PP54-100UG) One vial containing 100μg purified mouse IgG control antibody. Store at -20°C.
  • Rabbit IgG: (Catalog No. PP64-100UG) One vial containing 100μg purified rabbit IgG antibody. Store at -20°C.
When stored at the recommended storage conditions, components are stable up to the expiration date. Do not expose to elevated temperatures. Discard any remaining reagents after the expiration date.

For Research Use Only