missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Chemicon™ Human Embryonic Germ Layer Characterization Kit

Catalog No. SCR030
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SCR030 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SCR030 Supplier MilliporeSigma™ Supplier No. SCR030

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Allows researchers to assess differentiation potential of Human Embryonic Stem Cell (HESC) to form derivatives of all three embryonic germ layers. MilliporeSigma™; Chemicon™ Human Embryonic Germ Layer Characterization Kit is used in Immunocytochemistry, Stem cell culture applications.

  • Included in the kit are antibodies that are routinely used by stem cell researchers to identify the lineage fates of HESC following either spontaneous or directed differentiation:
  • Mesoderm: Smooth Muscle Actin (CBL171)
  • Cardiac Troponin I (MAB1691-50UG)
  • Endoderm: Alpha Fetoprotein (2004189)
  • Ectoderm: Nestin (ABD69)
  • MAP-2 (MAB3418-50UG)

Specifications

Cell Type Human Embryonic Stem Cells, Induced Pluripotent Stem Cells
Product Type Stem Cell Characterization Kit
For Use With (Application) Immunocytochemistry, Stem Cell Culture
Quantity 1 Kit
Regulatory Status RUO
Content And Storage
  • 50μL of Alpha Fetoprotein mouse monoclonal 2004189
  • 50μg of Smooth Muscle Actin mouse monoclonal CBL171
  • 50μL of Nestin rabbit polyclonal ABD69
  • 50μg of Cardiac Troponin I mouse monoclonal MAB1691-50UG
  • 50μg of MAP-2 mouse monoclonal MAB3418-50UG
Maintain components according to the individual data sheets. Antibodies to alpha fetoprotein (2004189) and nestin (ABD69) are to be stored at -20°C (SCR030 - Part 1). Antibodies to smooth muscle actin (CBL171), cardiac troponin I (MAB1691-50UG) and MAP-2 (MAB3418-50UG) are to be stored at 2° to 8°C (SCR030 - Part 2).

For Research Use Only