missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Chemicon™ ESGRO Complete™ Basal Medium

Catalog No. SF002100
Change view
Click to view available options
Quantity:
100 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SF002100 100 mL
SF002500 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. SF002100 Supplier MilliporeSigma™ Supplier No. SF002100

a defined serum-free medium developed for use as a base medium for differentiation studies, for serum free culture of ES cells when used with LIF and BMP.

ESGRO Complete™ Basal Medium is a defined serum-free medium developed for use as a base medium for differentiation studies, for serum free culture of ES cells when used with LIF and BMP, and during routine washing steps when passaging cells.

Specifications

For Use With (Application) Stem Cell Culture
Quantity 100 mL
pH 7.1 to 7.3
Content And Storage -20° C