missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Chemicon™ Embryonic Stem Cell Characterization Kit

Catalog No. SCR001
Change view
Click to view available options
Quantity:
100 Assays
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SCR001 100 Assays
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SCR001 Supplier MilliporeSigma™ Supplier No. SCR001

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Used in Immunocytochemistry applications. Chemicon™ Embryonic Stem Cell Characterization Kit is a specific and sensitive tool for phenotypic assessment of the differentiation status of ES cells by measuring AP activity, cell-surface stage-specific antigens (SSEA-1, SSEA-4), expression of TRA-1-60, TRA-1-81 antigens.

  • Consists of two components used for alkaline phosphatase activity determination as well as four ES cell-specific antibodies required to perform 100 tests (including controls)
  • Species Reactivity: Human, Mouse, Rat

Specifications

Cell Type Human Embryonic Stem Cells, Mouse Embryonic Stem Cells, Induced Pluripotent Stem Cells
Product Type Stem Cell Characterization Kit
For Use With (Application) Immunocytochemistry
Quantity 100 Assays
Regulatory Status RUO
Content And Storage
  • Fast Red Violet solution (0.8g/L stock) (Part No. 90239). One 15mL bottle.
  • Napthol AS-BI phosphate solution (4mg/mL) in AMPD buffer (2mol/L), pH 9.5 (Part No. 90234). One 15mL bottle.
  • MS X SSEA-1, IgM, clone MC-480 (Part No. 90230). One vial containing 100μl of 1mg/mL monoclonal antibody.
  • MS X SSEA-4, IgG, clone MC-813-70 (Part No. 90231). One vial containing 100μl of 1mg/mL monoclonal antibody.
  • MS X TRA-1-60, IgM, clone TRA-1-60 (Part No. 90232). One vial containing 100μl of 1mg/mL monoclonal antibody
  • MS X TRA-1-81, IgM, clone TRA-1-81 (Part No: 90233). One vial containing 100μl of 1mg/mL monoclonal antibody.
When stored at 2 to 8º C, the kit components are stable up to the expiration date. Do not freeze or expose to elevated temperatures. Discard any remaining reagents after the expiration date.

For Research Use Only