missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Chemicon™ Chicken Tenascin

Numéro de catalogue. CC115
Modifier l'affichage
Click to view available options
Quantité:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantité
CC115 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. CC115 Fournisseur MilliporeSigma™ Code fournisseur CC115

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

For positive control, cell culture applications

  • Large glycoprotein that forms disulfide-linked hexamers
  • Made up of repeated structural domains including EGF-like repeats, fibronectin type III repeats, and region homologous to fibrinogen
  • Binds to several cell-surface receptors and mediates cell-substrate adhesion
  • Frequently referred to as anti-adhesion molecule
  • Binding of tenascin to certain receptors inhibits cell spreading
  • Blocks stabilization of adhesion that results from cell preading in vitro assays

Spécifications

Concentration 0.755 mg/mL
À utiliser avec (application) Positive control, Cell Culture
Quantité 100 μg
Type de produit Chicken Tenascin
Contenu et stockage Store at -20°C for up to 12 months. Avoid repeated freeze/thaw.