missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Chemicon™ Adult Rat Neural Stem Cell Expansion Kit

Catalog No. SCR034
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SCR034 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SCR034 Supplier MilliporeSigma™ Supplier No. SCR034

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Used in Immunocytochemistry, Stem cell culture applications. MilliporeSigma™; Chemicon™ Adult Rat Neural Stem Cell Expansion Kit provides a multi-component system for the culture and analysis of neural stem cells and their differentiated progenies.

  • Includes primary neural stem cells isolated from the hippocampus of adult Fisher 344 rats, neural stem cell expansion medium, and a panel of antibodies for immunocytochemical staining of neural stem/progenitor cells (Nestin and Sox2) and differentiated neural phenotypes (Map2ab for neurons, GFAP for astrocytes, and O1 for oligodendrocytes)

Specifications

Cell Type Neural Stem Cells
Product Type Stem Cell Differentiation Media
For Use With (Application) Immunocytochemistry, Stem Cell Culture
Quantity 1 Kit
Regulatory Status RUO
Content And Storage
  • Adult Rat Hippocampal Neural Stem Cells (Cat.# SCR022)
  • Rat Neural Stem Cell Expansion Medium (Cat.# SCM009)
  • Neural Stem Cell Marker Characterization Kit (Cat.# SCR019)

For Research Use Only