missing translation for 'onlineSavingsMsg'
Learn More
Learn More
National Safety Apparel™ Chemical Resistant FR Lab Coats with Pockets
Navy chemical resistant lab coat made of flame-resistant CR/FR fabric for multi-hazard protection. Pockets feature covered flaps to secure contents.
Supplier: National Safety Apparel™ LABCRNBXL
Description
- Fabric/Material 4.4 oz. CR/FR Aramid Blend FR Nomex knit wrists keep sleeves in place
- Bottom side vents for increased mobility
- Durable hi-temp plastic snaps for easy donning and doffing
- Side pass-through allows access to pockets underneath the coat
- Durable accessory loop for safety glasses, pens, badges, etc.
- Chest pocket with covered flap and pen/pencil slot
- Lower pockets with covered flaps to secure contents
- Highly breathable
- Easy care and laundering
Specifications
| Chemical Resistant Lab Coat | |
| Unisex | |
| Snaps | |
| Reusable | |
| Chemicals, Flame | |
| ASTM F1506, NFPA 2112 |
| X-Large | |
| Navy | |
| Knit | |
| Non-sterile | |
| Knee-length |