missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CHEMetrics CHEMets Chlorine Test Kit Refill
Provides results in two minutes or less
$109.74 - $109.74
Specifications
| Analytes | Chlorine |
|---|---|
| For Use With (Equipment) | Chlorine (free and total) testing kit |
| Includes | 30 ampoules |
| Test Time | 2 min. |
| Type | CHEMets Refill |
| Catalog Number | Mfr. No. | Range | Test Method | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Range | Test Method | Price | Quantity | ||||
|
13-299-508B
|
CHEMetrics
R2500 |
0.1 to 2 ppm | DPD |
Pack of 30 Each for $109.74
|
|
||||
Description
- Designed for maximum simplicity and accuracy, Chemets 7 mm diameter ampules feature tapered, prescored tips with reagent vacuum-sealed inside
- To perform a test, simply immerse Chemets ampule in the sample, snap off tip and the correct sample volume is quickly drawn inside the ampule
- In two minutes or less, the reaction between the sample and reagent yields a color—which can then be compared to appropriate color standard for quantification of results
- Comparators have two-year shelf life
Specifications
| Chlorine | |
| 30 ampoules | |
| CHEMets Refill |
| Chlorine (free and total) testing kit | |
| 2 min. |