missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CHEMetrics Mercury-Free COD Vials Kits
Description
- No need to modify or recalibrate existing equipment or procedures
- Optical-quality glass for accuracy
- Available in mercury and mercury-free versions
- Standard 5/8 in. (16 mm) diameter for compatibility with standard COD reactors and spectrophotometers; can also be used in micro-COD reactors
- Follow the EPA-approved method for compliance reporting and use potassium dichromate as the oxidizing agent, silver sulfate as the catalyst, and mercury chloride to remove chloride interference
- Mercury-free version uses the same oxidizing agent and catalyst without the chloride compensator
Specifications
Specifications
| Analytes | Chemical Oxygen Demand (COD) |
| Description | A fresh reagent ampoule blank must be prepared for each series of tests; therefore the number of samples that can be tested with each kit will vary. |
| For Use With (Application) | Premeasured reagents for accurate, economical determination of COD, determination of the oxidizable organic matter content of wastewater |
| For Use With (Equipment) | Digestor block as well as a photometer or spectrophotometer |
| Includes | 25 vials and instruction book |
| No. of Tests | 24 |
| Range | 0 to 15,000 ppm (HR+) |
| Test Method | Dichromate Reactor Digestion |
| Test Time | 2 hr. digestion |
| Type | COD Vials Kit |