missing translation for 'onlineSavingsMsg'
Learn More

CHEMetrics™ Dissolved Oxygen Comparator

Color standard ampoules used to quantify test results for the determination of various analytes in water

Supplier:  CHEMetrics™ C7511DUP

Catalog No. 35-367

Only null left

Description

Description

  • Visual system
  • 0 to 20ppb
  • Shelf Life: 2 years

Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions