missing translation for 'onlineSavingsMsg'
Learn More

CHEMetrics COD Vials Kits

Catalog No. 361007032
Change view
Click to view available options
Range:
0 to 15,000 ppm (HR+)
0 to 150 ppm
0 to 1500 ppm
Certifications/Compliance:
Not USEPA approved
USEPA accepted
No. of Tests:
149
24
25
Includes:
150 vials and instruction book
25 vials and instruction book
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Range Certifications/Compliance No. of Tests Includes
361007032 0 to 150 ppm USEPA accepted 149 150 vials and instruction book
13299730 0 to 150 ppm USEPA accepted 24 25 vials and instruction book
13299732 0 to 1500 ppm USEPA accepted 25 25 vials and instruction book
13299230 0 to 1500 ppm USEPA accepted 149 150 vials and instruction book
13299437 0 to 15,000 ppm (HR+) Not USEPA approved 24 25 vials and instruction book
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 361007032 Supplier CHEMetrics Supplier No. K7355
Edge

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Includes

Instructions

Requires

Requires the use of a Digestor Block and the V-2000 Photometer, a COD Photometer, or a spectrophotometer capable of accepting a 16mm round cell. Instruments sold separately.

Contains premeasured reagents for accurate, economical determination of COD

  • Contains everything needed (except distilled water) to perform tests
  • Includes vials and instruction book
  • Comes in cardboard box
  • 2-hour digestion time
  • Dichromate reactor digestion

A fresh reagent ampule blank must be prepared for each series of tests; therefore the number of samples that can be tested with each kit will vary.

Certifications

USEPA-accepted

Specifications

Analytes Chemical Oxygen Demand (COD)
Certifications/Compliance USEPA accepted
For Use With (Application) Premeasured reagents for accurate, economical determination of COD, determination of the oxidizable organic matter content of wastewater
For Use With (Equipment) Digestor block as well as a photometer or spectrophotometer
Includes 150 vials and instruction book
No. of Tests 149
Range 0 to 150 ppm
Test Method Dichromate reactor digestion
Test Time 2 hr. digestion
Type COD Vials Kit