Learn More
CHEMetrics CHEMets Phosphate, ortho Visual Water Test Kit
Provides results in two minutes or less
Supplier: CHEMetrics K8510
Includes: REMOVE
Description
Provide results in two minutes or less
Kit comes in a plastic case and contains everything needed to perform 30 tests: Refill, Low & High Range Comparators, Activator Solution, 25 mL sample cup, sample cup top and instructionsSpecifications
| Phosphate, ortho | |
| 30 | |
| Stannous Chloride | |
| CHEMets Visual Water Testing Kit |
| Plastic case, Refill, Low and High range comparators, Activator solution, 25 mL sample cup, sample cup top instructions | |
| 0 to 1 ppm and 1 to 10 ppm | |
| 2 min. |