Learn More
CHEMetrics CHEMets Copper (Soluble) Visual Water Test Kit
Provides results in two minutes or less
Supplier:  CHEMetrics K3510
Catalog No. 13-299-216A
Only null left
Description
Description
Provide results in two minutes or less
Kit comes in a plastic case and contains everything needed to perform 30 tests: Refill, Low & High Range Comparators, 25 mL sample cup and instructions
Specifications
Specifications
Copper, Soluble
30
Bathocuproine
Plastic case, Refill, Low and High range comparators, 25 mL sample cup, instructions
0 to 1 ppm and 1 to 10 ppm
CHEMets Visual Water Test Kit
Product Suggestions
Product Suggestions