Learn More
CHEMetrics CHEMets Nitrate Visual Water Test Kit
Provides results in two minutes or less
Supplier:  CHEMetrics K6904
Catalog No. 13-299-252A
Only null left
Description
Description
Provide results in two minutes or less
Kit comes in a plastic case and contains everything needed to perform 30 tests: Refill, Comparator, 25 mL sample cup, reaction tube with lid and instructions.
Specifications
Specifications
Nitrate
30
Cadmium Reduction
CHEMets Visual Water Testing Kit
Plastic case, Refill, Comparator, 25 mL sample cup, reaction tube with lid and instructions
0 to 4.5 ppm as N
15 min.
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Promotions
Promotions
Warning! Prop 65