Learn More
CHEMetrics Sulfite (Free) Titrets Titration Cell Water Testing Kit
Provides results in two minutes or less
Supplier:  CHEMetrics K9605
Catalog No. 13-299-216X
Only null left
Explore available promotions
Includes: REMOVE
Description
Description
Provide results in two minutes or less
Kit comes in a cardboard box and contains everything needed to perform 30 sulfite titration tests: 30 ampules with valve assemblies, Neutralizer Solution, titrettor, 25 mL sample cup and instructions
Specifications
Specifications
Sulfite, Free
30
Iodometric
Cardboard box, 30 ampoules with valve assemblies, titrettor, 25 mL sample cup, instructions, Neutralizer solution
5 to 50 ppm as SO3
Titrets Titration Cell Water Testing Kit
Product Suggestions
Product Suggestions