Learn More
CHEMetrics CHEMets Copper Test Kit Refill
Provides results in two minutes or less
Supplier: CHEMetrics R3510
Description
This product contains 30 self-filling CHEMets ampules to refill kit K-3510 for the analysis of copper (soluble). R-3510 is compatible with C-3501 and C-3510 comparators.Specifications
| Copper | |
| 30 self-filling CHEMets ampoules to refill kit K-3510 | |
| 0 to 1 ppm and 1 to 10 ppm | |
| 2 min. |
| C-3501 and C-3510 comparators | |
| 30 | |
| Bathocuproine | |
| CHEMets Refill Kit |