Learn More
CHEMetrics CHEMets Phenol Test Kit Refill
Catalog No. 13299205A
Change view
Click to view available options
No. of Tests:
30
Catalog No. No. of Tests
13299205A 30
1 options
Catalog No. 13299205A Supplier CHEMetrics Supplier No. R8012
Edge
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Provides results in two minutes or less
This product contains 30 self-filling CHEMets ampules to refill kit K-8012 for the analysis of phenols. R-8012 is compatible with C-8001 and C-8012 comparators.
Specifications
Analytes Phenol
For Use With (Equipment) C-8001 and C-8012 comparators
Includes 30 self-filling CHEMets ampoules to refill kit K-8012
No. of Tests 30
Range 0 to 1 ppm and 1 to 12 ppm
Test Method 4-Aminoantipyrine
Type CHEMets Refill Kit