missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CHEMetrics CHEMets Dissolved Oxygen Test Kit Refill
Provides results in two minutes or less
$126.74 - $164.03
Specifications
| Analytes | Dissolved Oxygen (DO) |
|---|---|
| No. of Tests | 30 |
| Test Time | 2 min. |
| Type | CHEMets Dissolved Oxygen Refill |
| Catalog Number | Mfr. No. | Range | Test Method | For Use With (Equipment) | Price | Quantity | |||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Range | Test Method | For Use With (Equipment) | Price | Quantity | |||
|
13-299-201A
|
CHEMetrics
R7501 |
0 to 1 ppm | Rhodazine D | C-7501 comparators |
Pack of 30 Each for $126.74
|
|
|||
|
13-299-200B
|
CHEMetrics
R7512 |
1 to 12 ppm | Indigo Carmine | C-7512 comparators |
Pack of 30 Each for $126.74
|
|
|||
|
13-299-202A
|
CHEMetrics
R7540 |
0 to 40 ppb and 0 to 100 ppb | Rhodazine D | C-7540 comparators |
Pack of 30 Each for $164.03
|
|
|||
Description
- Designed for maximum simplicity and accuracy, Chemets 7 mm diameter ampules feature tapered, prescored tips with reagent vacuum-sealed inside
- To perform a test, simply immerse Chemets ampule in the sample, snap off tip and the correct sample volume is quickly drawn inside the ampule
- In two minutes or less, the reaction between the sample and reagent yields a color—which can then be compared to appropriate color standard for quantification of results
- Comparators have two-year shelf life
Specifications
| Dissolved Oxygen (DO) | |
| 30 | |
| 2 min. | |
| CHEMets Dissolved Oxygen Refill |