Please login to your online account to display your discounted pricing

CHEMetrics™ Chemets™ Test Kit Refills


Provide results in two minutes or less

Manufacturer: chemetrics™  CHIR9400

 View more versions of this product

Catalog No. 13-299-261B

  • / Pack of 20 Each
 

Description & Specifications

Specifications

For Use With (Equipment) Detergents testing kits
Includes 30 ampoules
Shelf Life 8 Months
Method Methylene Blue
Product Type Chemets™ Refill
Range 0 to 3ppm