missing translation for 'onlineSavingsMsg'
Learn More

Chelidamic Acid Monohydrate 95.0+%, TCI America™

Catalog No. p-7124628
Change view
Click to view available options
Quantity:
5 g
25 g
Catalog No. Quantity
C08215G 5 g
C082125G 25 g
2 options

Catalog No. C08215G

Supplier: TCI America C08215G

Only null left

Chemical Identifiers

CAS 138-60-3
Molecular Formula C7H5NO5
Molecular Weight (g/mol) 183.12
MDL Number MFCD00150943
InChI Key XTLJJHGQACAZMS-UHFFFAOYSA-N
Synonym chelidamic acid, 4-hydroxypyridine-2,6-dicarboxylic acid, 4-oxo-1,4-dihydropyridine-2,6-dicarboxylic acid, 4-hydroxy-2,6-pyridinedicarboxylic acid, chelidamic acid hydrate, 2,6-pyridinedicarboxylic acid, 1,4-dihydro-4-oxo, 2,6-dicarboxy-4-hydroxypyridine, 1,4-dihydro-4-oxopyridine-2,6-dicarboxylic acid, 2,6-pyridinedicarboxylic acid, 4-hydroxy, 4-oxo-1,4-dihydro-2,6-pyridinedicarboxylic acid
PubChem CID 8743
IUPAC Name 4-oxo-1,4-dihydropyridine-2,6-dicarboxylic acid
SMILES OC(=O)C1=CC(=O)C=C(N1)C(O)=O

Specifications

Melting Point 267°C
Color White-Yellow
Quantity 5 g
Formula Weight 183.12
Percent Purity ≥95.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Chelidamic Acid Monohydrate