missing translation for 'onlineSavingsMsg'
Learn More

Chelidamic acid hydrate, 97%, Thermo Scientific™

Catalog No. p-7077761
Change view
Click to view available options
Quantity:
1g
5g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC458870010 1g
AC458870050 5g
AC458870250 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC458870010 Supplier Thermo Scientific Chemicals Supplier No. 458870010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Chemical Identifiers

CAS 138-60-3
Molecular Formula C7H5NO5·xH2O
Molecular Weight (g/mol) 183.12
InChI Key XTLJJHGQACAZMS-UHFFFAOYSA-N
Synonym chelidamic acid, 4-hydroxypyridine-2,6-dicarboxylic acid, 4-oxo-1,4-dihydropyridine-2,6-dicarboxylic acid, 4-hydroxy-2,6-pyridinedicarboxylic acid, chelidamic acid hydrate, 2,6-pyridinedicarboxylic acid, 1,4-dihydro-4-oxo, 2,6-dicarboxy-4-hydroxypyridine, 1,4-dihydro-4-oxopyridine-2,6-dicarboxylic acid, 2,6-pyridinedicarboxylic acid, 4-hydroxy, 4-oxo-1,4-dihydro-2,6-pyridinedicarboxylic acid
PubChem CID 8743
IUPAC Name 4-oxo-1H-pyridine-2,6-dicarboxylic acid
SMILES C1=C(NC(=CC1=O)C(=O)O)C(=O)O

Specifications

Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass Bottle
Quantity 1g
Formula Weight 183.12
Percent Purity 97%
Chemical Name or Material Chelidamic acid hydrate
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and kee
EINECSNumber 205-335-8

RUO – Research Use Only