missing translation for 'onlineSavingsMsg'
Learn More

Chelidamic acid hydrate, 95%

Catalog No. AAL0078203
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0078203 1 g
AAL0078206 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0078203 Supplier Thermo Scientific Chemicals Supplier No. L0078203

CAS: 138-60-3 | C7H5NO5 | 183.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 138-60-3
Molecular Formula C7H5NO5
Molecular Weight (g/mol) 183.12
MDL Number MFCD00150943
InChI Key XTLJJHGQACAZMS-UHFFFAOYSA-N
Synonym chelidamic acid, 4-hydroxypyridine-2,6-dicarboxylic acid, 4-oxo-1,4-dihydropyridine-2,6-dicarboxylic acid, 4-hydroxy-2,6-pyridinedicarboxylic acid, chelidamic acid hydrate, 2,6-pyridinedicarboxylic acid, 1,4-dihydro-4-oxo, 2,6-dicarboxy-4-hydroxypyridine, 1,4-dihydro-4-oxopyridine-2,6-dicarboxylic acid, 2,6-pyridinedicarboxylic acid, 4-hydroxy, 4-oxo-1,4-dihydro-2,6-pyridinedicarboxylic acid
PubChem CID 8743
IUPAC Name 4-oxo-1,4-dihydropyridine-2,6-dicarboxylic acid
SMILES OC(=O)C1=CC(=O)C=C(N1)C(O)=O

Specifications

Melting Point ∽260°C (decomposition)
Quantity 1 g
Formula Weight 183.12 (Anhydrous)
Percent Purity 95%
Chemical Name or Material Chelidamic acid hydrate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 205-335-8
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only