Learn More
Thermo Scientific™ Chart Recorders for Ultra-Low Temperature Freezers
Record seven days of temperature performance at temperatures to -150° C.
Supplier:  Thermo Scientific™ FCR400TSXLE
 View more versions of this product
Catalog No. FCR400TSXLE
Only null left
Description
Description
Record seven days of temperature performance at temperatures to -150° C using Thermo Scientific™ Chart Recorders for Ultra-Low Temperature Freezers. Recorders use 6 in. (15cm) chart paper and are available as factory or field installed.
Field-installed ink chart recorder
Warranty
Warranty varies by country, please contact us for details
Specifications
Chemical Identifiers
Recorder
Specifications
Inkless Chart Recorder, Field Installed
TSX400 Ultra-low Freezers
Recorder
Product Suggestions
Product Suggestions