Learn More
Thermo Scientific™ Chart Recorders for Ultra-Low Temperature Freezers
Record seven days of temperature performance at temperatures to -150° C.
Supplier:  Thermo Scientific™ FCR400LE
 View more versions of this product
Catalog No. FCR400LE
Only null left
Description
Description
Record seven days of temperature performance at temperatures to -150° C using Thermo Scientific™ Chart Recorders for Ultra-Low Temperature Freezers. Recorders use 6 in. (15cm) chart paper and are available as factory or field installed.
Field-installed ink chart recorder
Warranty
Warranty varies by country, please contact us for details
Specifications
Chemical Identifiers
Recorder
Specifications
Inkless Chart Recorder, Field Installed
Revco RLE, Forma 89000, TLE Series, HLE Series (400 models only)
Recorder
Product Suggestions
Product Suggestions