Learn More
Eppendorf™ Charger Stand 2
Description
- Carries and charges one Multipette™ E3/E3x or Multipette™ stream/Xstream
- Robust design that securely holds and protects the instrument
- Large rubber feet protect stand from liquids spilled on workbench
- Cord wrap functionality for unused power cord
- Operated with mains/power adapter supplied with Multipette E3/E3x or Multipette stream/Xstream
Specifications
Specifications
| Holds | Multipette™ E3/E3x or Multipette™ stream/Xstream |
| For Use With (Equipment) | One Multipette™ E3/E3x or Multipette™ stream/Xstream, operated with mains/power adapter supplied with Multipette™ E3/E3x or Multipette™ stream/Xstream |
| Product Type | Pipette Holder and Charger |