Learn More
Eppendorf™ Charger Carousel 2
GREENER_CHOICE
Catalog No. 13690230
Change view
Click to view available options
Positions:
6
Catalog No. Positions
13690230 6
1 options
Catalog No. 13690230 Supplier Eppendorf™ Supplier No. 3116000023
Edge
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Carries and charges up to six Eppendorf Xplorer™ or Xplorer plus™ models.
  • When used with optional charger shells, the carousel can carry and charge Multipette E3/E3x and Multipette stream/Xstream
  • Also perfect for manual Eppendorf pipettes when used with optional pipette holders
  • Pipette holders and charger shells are interchangeable, no tools required
  • Handle for easy transport from bench to bench
  • Magnetic power connector for fast and easy connection
  • Intelligent charging electronics with small power adapter
  • Large rubber feet protect carousels from liquids spilled on workbench
  • Cord wrap functionality for unused power cord
  • Includes four interchangeable power plugs for different countries, 100 – 240 Volts, 50/60 Hz
Specifications
Product Type Pippetter Charging Stand
Device Holds Up to six Eppendorf Xplorer™ or Xplorer plus™ models
For Use With (Equipment) 6 Eppendorf Xplorer™ or Xplorer™ plus Pipetters
Model Charger Carousel 2
Positions 6
Electrical Requirements 100 - 240 V, 50/60 Hz